Learn More
BD BBL™ Dehydrated Culture Media: Litmus Milk
For the maintenance of lactic acid bacteria
Brand:  BD 211343
Additional Details : CAS Number : 7757-83-7 Weight : 0.50000kg
Product Code. 11728123
  • 3325.00 NOK / 500g
Estimated Shipment: 07-08-2024
Description
  • Used as differential medium for determining the action of bacteria on milk
  • Determines metabolic activities of microorganisms in milk as aid to identification of bacterial species
  • Useful in species differentiation within genus Clostridium
  • Appearance: Fine, homogeneous, free of extraneous material
  • Solution: 10% solution, soluble in purified water. Solution is medium, purple gray, opaque
Applications:
  • For evaluation, maintenance, selective enrichment, differentiation, isolation, cultivation, detection, propagation of bacteria
Prepare the medium per label directions. Inoculate with fresh cultures diluted 1:10 and incubate at 35±2°C for 7 days.
Specifications
Specifications
Dehydrated
Dehydrated Culture Media
Used for the maintenance of lactic Acid bacteria and as a differential medium for determining the action of bacteria on Milk
500 g
Litmus Milk
Skim Milk 100g/L, Azolitmin 0.5g/L, Sodium Sulfite 0.5g/L
Bottle