missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: DNase Test Agar (Deoxyribonuclease Test)
Differentiates microorganisms based on DNase activity
Brand: BD 263220
This item is not returnable.
View return policy
Description

Specifications
COMPF | |
Dehydrated | |
Agar | |
For differentiating microorganisms based on DNase activity | |
500 g |
Refrigerate | |
DNase Test Agar | |
Tryptose 20g/L, Deoxyribonucleic Acid 2g/L, Sodium Chloride 5g/L, Agar 15g/L | |
Bottle |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BD Difco™ Dehydrated Culture Media: DNase Test Agar (Deoxyribonuclease Test) > DNase Test Agar (Deoxyribonuclease Test); 500g
Spot an opportunity for improvement?Share a Content Correction