Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Ticarcillin
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents
Brand:  BD 231619
Product Code. 17332391
  • 2450.00 NOK / Pack of 10
Estimated Shipment: 25-02-2025
to see stock.
Explore more special offers
Description
  • Clearly marked on both sides with letters and numbers designating agent and concentration
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ticarcillin
75μg
Disc
10/Pk.