missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCL10 Mouse anti-Human, Clone: 2E8G5, Proteintech
Mouse Monoclonal Antibody
1696.32 NOK - 4796.50 NOK
Specifications
Antigen | BCL10 |
---|---|
Clone | 2E8G5 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16808545
|
Proteintech
66556-1-IG-20UL |
20 μL |
1696.32 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16898535
|
Proteintech
66556-1-IG-150UL |
150 μL |
4796.50 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BCL-10 is an N-terminal CARD (Caspase Recruitment Domain) containing protein that is involved in the adaptive immune response. It promotes apoptosis, pro-caspase-9 maturation, and activation of NF-kappa-B via NIK and IKK. BCL-10 is also an adapter protein between upstream TNFR1-TRADD-RIP complex and the downstream NIK-IKK-IKAP complex. It is a substrate for MALT1. Mutations in the gene can lead to Immmunodeficiency 37 or Lymphoma, mucosa-associated lymphoid type.Specifications
BCL10 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
B cell CLL/lymphoma 10, B cell lymphoma/leukemia 10, BCL10, c E10, CARD like apoptotic protein, CARMEN, cCARMEN, Cellular E10, Cellular homolog of vCARMEN, CIPER, CLAP, hCLAP, mE10 | |
BCL10 | |
IgG1 | |
Protein G | |
Antibody |
2E8G5 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
O95999 | |
8915 | |
BCL10 Fusion Protein Ag12162 | |
Primary | |
-20°C | |
BCL10 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title