Learn More
Pierce™ BCA Solid
Brand: Thermo Scientific 23230
2658.75 NOK valid until 2024-12-31
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
C20H10N2Na2O4 | |
25 g | |
[Na+].[Na+].[O-]C(=O)C1=C2C=CC=CC2=NC(=C1)C1=CC(C([O-])=O)=C2C=CC=CC2=N1 | |
388.29 | |
Pierce |
MFCD00037500 | |
AUPXFICLXPLHBB-UHFFFAOYSA-L | |
disodium [2,2'-biquinoline]-4,4'-dicarboxylate | |
Spectrophotometer, Microplate Reader |