Learn More
Brady™ BBP™ 33 Series WorkHorse™ Chemical Resistant, Extreme Temperature Polypropylene Labels
BBP™ 33 Series WorkHorse™ Chemical Resistant, Extreme Temperature Polypropylene Labels are ideal for general purpose barcode applications and withstand liquid nitrogen, autoclave and other harsh environments.
Brand:  Brady™ 148672
 View more versions of this product
Product Code. 17160171
  • 1760.00 NOK / Pack of 1250
Estimated Shipment: 09-05-2025
to see stock.
Explore more special offers
Description
  • Perfect for laboratory identification such as vials, centrifuge tubes and test tubes
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
  • Service temperature of -112 to 158° F (-80 to 70° C)
Compliance
ASTM D 1000;ASTM D 2979;RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
Specifications
Specifications
B33-235-7425
White
Acrylic
BBP™33 Label Printer
1.5 in.
Acrylic
Matte
Thermal Transfer
1250/Pk.
1 in.
Documents
One moment while we fetch your results.
Product Title
Brady™ BBP™ 33 Series WorkHorse™ Chemical Resistant, Extreme Temperature Polypropylene Labels > Height: 1in; Width: 1.5in; Quantity: 1250