Learn More
BD BBL™ CultureSwab EZ™ and CultureSwab EZ II™
A patented media-free collection system designed to collect aerobic specimens and maintain viability during transport
1005.00 NOK
Specifications
Sterility | Sterile |
---|---|
For Use With (Application) | For collecting and transporting aerobic microbiological specimens |
Quantity | 100/Pk. |
Shaft Material | Plastic |
Storage Requirements | 15°C to 30°C |
Product Code | Brand | Includes | Product Type | Swab Configuration | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Product Type | Swab Configuration | Price | Quantity & Availability | |||
14871121
|
BD
220144 |
Polyurethane single sponge swab, Plastic shaft, cap, inserted into Plastic transport tube | Culture Swab™ EZ System | Single Applicator | N/A | ||||
Description
- No specimen dilution with this dry system and no ampule to crush
- Transport system eliminates the organism overgrowth and nonviables typically present in agar-based systems
- Aerobic microorganisms remain viable for approximately 48 hours
- Features rounded tube bottom for easier handling, and water-resistant film packing with tamper-evident seal
- BD CultureSwab EZ II is similar to BD CultureSwab EZ except formatted as a dual swab system, allowing for simultaneous gram staining and other direct antigen testing
Specifications
Sterile | |
100/Pk. | |
15°C to 30°C |
For collecting and transporting aerobic microbiological specimens | |
Plastic |