Learn More
Emma Safety Footwear Barocco (Frontier 168) Safety Shoes
Brazilian 2-tone full grain leather combined with nubuck adds to the stylish look.
2400.00 NOK - 2730.00 NOK
Specifications
Certifications/Compliance | EN ISO 20345:2011, EN 12568:2010 |
---|---|
Closure Type | Laces |
Color | Blue |
Design | Barocco (Frontier 168) |
Liner Material | Hydro-Tec™ Sanitized Silver & Wellness leather |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
15880740
|
Emma Safety Footwear
MM16809039 |
39 |
2400.00 NOK
Each |
Estimated Shipment: 21-08-2025
Log in to see stock. |
||||||
15890740
|
Emma Safety Footwear
MM16809040 |
40 |
2455.00 NOK
Each |
Estimated Shipment: 21-08-2025
Log in to see stock. |
||||||
15800750
|
Emma Safety Footwear
MM16809041 |
41 |
2400.00 NOK
1 pair |
Estimated Shipment: 21-08-2025
Log in to see stock. |
||||||
15810750
|
Emma Safety Footwear
MM16809042 |
42 |
2455.00 NOK
Each |
Estimated Shipment: 21-08-2025
Log in to see stock. |
||||||
15820750
|
Emma Safety Footwear
MM16809043 |
43 |
2400.00 NOK
Each |
Estimated Shipment: 21-08-2025
Log in to see stock. |
||||||
15830750
|
Emma Safety Footwear
MM16809044 |
44 |
2730.00 NOK
Each |
Estimated Shipment: 21-08-2025
Log in to see stock. |
||||||
15840750
|
Emma Safety Footwear
MM16809045 |
45 |
2400.00 NOK
Each |
Estimated Shipment: 21-08-2025
Log in to see stock. |
||||||
15850750
|
Emma Safety Footwear
MM16809047 |
47 |
2400.00 NOK
Each |
Estimated Shipment: 21-08-2025
Log in to see stock. |
||||||
15860750
|
Emma Safety Footwear
MM16809048 |
48 |
2400.00 NOK
Each |
Estimated Shipment: 21-08-2025
Log in to see stock. |
||||||
Description
- S3-Safety shoe with sole plate for wet work
- TPU Outsole
- Full grain leather
- ESD Certified
- Chrome-free lining
- Hydro-Tec™ Sanitized Silver
Specifications
EN ISO 20345:2011, EN 12568:2010 | |
Blue | |
Hydro-Tec™ Sanitized Silver & Wellness leather | |
Steel |
Laces | |
Barocco (Frontier 168) | |
Leather |
Product Title
Emma Safety Footwear Barocco (Frontier 168) Safety Shoes