Learn More
Barbituric Acid, 99+%, Extra Pure, SLR, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
:
100g, Plastic powder jar
Specifications
Specifications
Melting Point | 248°C |
Color | Cream |
Quantity | 100 g |
Packaging | Plastic powder jar |
Product Content Correction
Product Title
Barbituric Acid, 99+%, Extra Pure, SLR, Fisher Chemical™ >