Learn More
JULABO™ Barbed Fittings
Barbed Fittings are fluid tubing adapters to convert male barbed fittings of various Internal Diameters (ID) to male G thread fittings with the same ID.
Brand:  JULABO™ 8890151
 View more versions of this product
Product Code. 17118508
  • 469.00 NOK / Each
Explore more special offers
Description
Description
  • Fitting styles include:
    • 3/4 in. ID male to G 3/4 in. male
    • 1 in. ID male to G 1 in. male
    • 1 1/2 in. ID male to G 1 1/2 in. male
Specifications
Specifications
1 in. ID Male to G 1 in. Male
Fitting Adapter, Fluid Tubing
2 Pk.