Learn More
BAG3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10599-1-AP-20UL
 View more versions of this product
Product Code. 16857083
  • 1696.32 NOK / 20µL
Explore more special offers
Description
Description
BAG3 are among BAG proteins that compete with Hip for binding to the Hsc70/Hsp70 ATPase domain and promote substrate release. All the BAG proteins have an approximately 45-amino acid BAG domain near the C terminus but differ markedly in their N-terminal regions. The protein encoded by this gene contains a WW domain in the N-terminal region and a BAG domain in the C-terminal region. The BAG domains of BAG1, BAG2, and BAG3 interact specifically with the Hsc70 ATPase domain in vitro and in mammalian cells. All 3 proteins bind with high affinity to the ATPase domain of Hsc70 and inhibit its chaperone activity in a Hip-repressible manner.
Specifications
Specifications
BAG3
Polyclonal
Unconjugated
BAG3
BAG 3, BAG3, BIS, CAIR 1, Docking protein CAIR 1
Rabbit
Antigen Affinity Chromatography
RUO
293524, 29810, 9531
-20°C
Liquid
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry
0.48 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O95817, Q9JLV1
BAG3
BAG3 Fusion Protein Ag0956
20 μL
Primary
Rat, Mouse, Human