Learn More
Steris™ Bag-in-Box
Brand:  Steris™ 1D20BGEC
Product Code. 15307023
  • 2305.00 NOK / Each
Only null left
Estimated Shipment: 12-11-2025
Specifications
Bag-in-Box
3.78 L
Documents
One moment while we fetch your results.
Product Certifications
Promotions
Promotions
Product Content Correction
Product Title
Steris™ Bag-in-Box > Capacity: 3.78L