missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAF53A Rabbit anti-Human, Mouse, Rat, Clone: 5U6J8, Novus Biologicals™
Rabbit Monoclonal Antibody
1815.00 NOK - 4580.00 NOK
Specifications
Antigen | BAF53A |
---|---|
Clone | 5U6J8 |
Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18329195
|
Bio-Techne
NBP3-16659-20UL |
20 μg |
1815.00 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18380533
|
Bio-Techne
NBP3-16659-100UL |
100 μg |
4580.00 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BAF53A Monoclonal antibody specifically detects BAF53A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
BAF53A | |
Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
actin-like 6A, actin-like protein 6A, actin-related protein 4,53 kDa BRG1-associated factor A, Actin-related protein Baf53a, Actl6, Arp4, ArpNbeta, BAF complex 53 kDa subunit, Baf53a, BAF53AACTL6, BAF53MGC5382, BRG1-associated factor 53A, hArpN beta, INO80 complex subunit K, INO80KARPN-BETA | |
Recombinant fusion protein containing a sequence corresponding to amino acids 201-290 of human BAF53A (O96019). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
5U6J8 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Extracellular Matrix | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
86 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title