missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Bacto™ Neopeptone

An animal origin (AO) enzymatic digest of bovine (milk) and porcine protein. This nutritional supplement provides vitamins, nucleotides, minerals, and peptides in a variety of sizes to any culture formulation.

Brand:  Gibco™ 211680

 View more versions of this product

Product Code. 16209751

  • 86100.00 NOK / 10kg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Bacto Neopeptone is an animal origin (AO) enzymatic digest of bovine (milk) and porcine protein
  • This nutritional supplement provides vitamins, nucleotides, minerals, and peptides in a variety of sizes to any culture formulation
  • It has been used as a component of culture media in the cultivation of human pathogens, notably Bordetella pertussis and group A streptococci
  • Also used with Borrelia burgdorferi and Clostridium species, to produce vaccines for animal health application
  • Store in cool, dry conditions in well-sealed containers at 2°C to 25°C

Applications of Bacto Neopeptone include:

  • Recommended for use in media for detection of fungi
  • Reported to provide nutrients for support of spirochetes and protozoa
  • Microbial fermentation
  • Production of vaccines for animal health applications
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Dehydrated Culture Media
Bacto™
Animal origin, bovine and porcine
Powder
Production of vaccines for animal health applications
10 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number4261485 Certificate TypeCertificate of Analysis Date24/10/2024 Catalog Number211680
Lot Number4074752 Certificate TypeCertificate of Analysis Date03/06/2024 Catalog Number211680
Lot Number3338176 Certificate TypeCertificate of Analysis Date02/01/2024 Catalog Number211680
Lot Number3205398 Certificate TypeCertificate of Analysis Date07/08/2023 Catalog Number211680
Lot Number9338690 Certificate TypeCertificate of Analysis Date15/06/2023 Catalog Number211680
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ Bacto™ Neopeptone > 10 kg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.