missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scharlab Bacillus Cereus Agar (According to Mossel Formula)
Scharlab Bacillus Cereus Agar is used for the isolation and identification of Bacillus cereus from food samples
Brand: Scharlab 01-262-500
This item is not returnable.
View return policy
Description
- COMPF/FIL-IDF/ISO
- Selective solid medium, according to Mossel, for the isolation and identification of Bacillus cereus from food samples.
- According to ISO standards 7932 and 21871.
- This medium needs add 100 mL of Egg Yolk Sterile Emulsion (Art. No. 06-016) and 100 mg/L of Polymyxin B (Art. No. 06-021LYO1).
Specifications
Bacillus cereus | |
Red | |
Scharlab Bacillus Cereus Agar | |
Gram-positive Organisms | |
Isolation and Identification of Bacillus Cereus From Food Samples | |
24 ±3 hr./44 ±4 hr. | |
500 g |
ISO 7932, 21871 | |
Solid | |
Microbiology Media | |
Peptone 10.000 g/L, Mannitol 10.000 g/L, Sodium Chloride 10.000 g/L, Meat Extract 1.000 g/L, Phenol Red 0.025 g/L, Agar 15.000 g/L | |
30°C ± 1°C | |
7.2 ±0.2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Scharlab Bacillus Cereus Agar (According to Mossel Formula) > Bacillus Cereus Agar; Form: Solid
Spot an opportunity for improvement?Share a Content Correction