missing translation for 'onlineSavingsMsg'
Learn More

Brady™ B-7610 Paper Thermal Transfer Labels, 25 mm Core

Thermal transfer printable paper for low cost general labeling applications.

Brand:  Brady™ 800843

 View more versions of this product

Product Code. 11548764

  • 1820.00 NOK / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 25 mm core
  • Permanent acrylic adhesive
  • Matte finish
Specifications

Specifications

Not Autoclavable
Permanent Acrylic
Matte
Thermal Transfer
Hazard Communication Identification
1
Rectangle
0.081 mm
55.9 mm
BPT LABELS 104 X 56 MM IN B-7610
White
Paper (B-7610)
Non-sterile
BBP™11, BBP™12, BradyPrinter i7100, PR Plus Series, and IP™ Series Printers, THT 76 mm Core Printers
4000 Labels
-40°C to +50°C
104 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.