Learn More
Azlon™ Magnetic Retrievers for Stirrer Bars
Retrieve stirrer bars from sensitive liquids with these magnetic retrievers. Azlon™ Magnetic Retrievers for Stirrer Bars feature magnets permanently embedded in a PTFE rod.
263.00 NOK - 384.00 NOK
Specifications
Autoclavable | Autoclavable |
---|---|
Coating | PTFE |
Diameter (Metric) | 10 mm |
Material | Magnet |
Quantity | 1/Pk. |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | ||||||
10514355
|
Azlon™
SWN564 |
250 mm |
263.00 NOK
Each |
Estimated Shipment: 28-07-2025
Log in to see stock. |
||||||
10400095
|
Azlon™
SWN566 |
350 mm |
384.00 NOK
Each |
Estimated Shipment: 24-07-2025
Log in to see stock. |
||||||
Description
Retrieve stirrer bars from sensitive liquids with these magnetic retrievers. AzlonTM Magnetic Retrievers for Stirrer Bars feature magnets permanently embedded in a PTFE rod.
- Ideal for retrieving magnetic stirrer bars from particularly sensitive or corrosive liquids
- Turbo retrievers are ideal for use with turbo stirrer bars
Specifications
Autoclavable | |
10 mm | |
1/Pk. |
PTFE | |
Magnet |