missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azlon™ Bottle Wide Neck 150mL 32mm HDPE Bulk Pack
HDPE bottles suitable for food use applications
Brand: Azlon BLH0150BULK
This item is not returnable.
View return policy
Description
This product is suitable for food contact applications in accordance with EU and\or USA regulations
Specifications
150 mL | |
HDPE |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Azlon™ Bottle Wide Neck 150mL 32mm HDPE Bulk Pack >
Spot an opportunity for improvement?Share a Content Correction