Learn More
Tocris Bioscience™ AZD 3988
Potent and selective DGAT-1 inhibitor
4810.00 NOK - 18865.00 NOK
Specifications
Purity | >98% |
---|---|
Product Type | AZD 3988 |
Molecular Weight (g/mol) | 456.44 |
Inhibitors | AZD 3988 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18716638
|
Tocris Bioscience™
4837/10 |
10 mg |
4810.00 NOK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18774988
|
Tocris Bioscience™
4837/50 |
50 mg |
18865.00 NOK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potent and selective diacylglycerol acyltransferase (DGAT-1) inhibitor (IC50 = 0.6 nM). Exhibits selectivity for DGAT-1 over DGAT-2, Kv11.1 (hERG) and cytochrome P450 enzymes. Suppresses triacylglyceride (TAG) plasma excursion and adipose tissue TAG synthesis in rats. Reduces body weight of diet-induced obese rats. Cell permeable and orally bioavailable.Specifications
>98% | |
AZD 3988 | |
456.44 | |
AZD 3988 |
For Research Use Only