Learn More
Axygen™ Axypet™ Starter Kit
CORNING AXYGEN™ Axypet™ Starter Kit includes a set of micropipettes, tips, and other quality accessories.
Brand: Axygen™ AP-STR-KIT-1
4383.99 NOK valid until 2025-06-30
Use promo code "24075" to get your promotional price.
Description
- Micropipette starter kit
- Includes micropipettes, tips, calibration tools, identification tools, and more
Specifications
Axypet | |
20 to 1000 μL | |
1 Kit |
Starter Kit, Micro | |
AP-20, AP-200, AP-1000 micropipettes, 1 rack each of 200μL and 1000μL tips, a calibration tool, ejector spacers, identification labels, lubricant, color identification rings, right-hand ejector button |
Promotions
-
Multiple Manufacturers
The Ultimate Lab Appliances Promotion
Enjoy discounts up to 78% on top-quality lab equipment and upgrade your laboratory with the best appliances at the lowest prices.Promo Code: 24075Expires: 2025-06-30
Days
11Hours
10Minutes
5Seconds
14