Learn More
Axygen™ Tip Refill System
Axygen™ style and universal fit pipet tips in easy-to-load refill racks
569.55 NOK - 775.58 NOK
Specifications
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
View More Specs
Products 3
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
11300285
View Documents Promotion Details
Axygen™
RFL-1200-C
15 to 1250 μL
775.58 NOK
Pack of 768
Estimated Shipment: 23-06-2025
to see stock.
Add to basket
11310285
View Documents Promotion Details
Axygen™
RFL-222-C
1 to 200 μL
569.55 NOK
Pack of 960
In Stock
to see stock.
Edge
Add to basket
11320285
View Documents Promotion Details
Axygen™
RFL-300-C
0.1 to 10 μL
687.68 NOK
Pack of 960
In Stock
to see stock.
Edge
Add to basket
Description
Description
Axygen™ Tip Refill System contains durable refill rack that can be easily refilled by hand and is autoclavable. These translucent Racks includes color-coded inserts for tip identification and are certified RNase-/DNase free. They are available in different colors and are packaged in bulk or racks.
  • Certified to be RNase-/DNase-free
  • Nonpyrogenic
  • Translucent rack allows tips to be easily seen
  • Color-coded insert allows for easy identification of tip
  • Specifications
    Specifications
    Non-sterile
    Clear
    Autoclavable
    Documents
    Product Certifications
    Product Content Correction
    Product Title
    Axygen™ Tip Refill System