Learn More
Axygen™ Presterilized Conical Centrifuge Tubes
GREENER_CHOICE
Graduated tubes with conical bottom
Brand:  Axygen™ SCT-50ML-25-S
 View more versions of this product
Product Code. 11344205
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Rated to 12,000 RCF
  • Printed graduations and writing area for sample identification
  • Screw caps can be closed with one hand.
  • Sterile options are also available.
  • RNase-/DNase-free
  • Nonpyrogenic
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Screw Cap
50 mL
Polypropylene
Screw cap
Sterile
12,000 x g
Not Autoclavable
Clear
For sample identification
25 Tubes and Caps/Unit, 20 Units/Cs.
Conical
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Axygen™ Presterilized Conical Centrifuge Tubes > 50 mL; Yes; SAL 10-5