Learn More
Axygen™ Axypet™ Pro Pipettor Starter Kit
Corning™ Axygen™ Axypet™ Pro Pipettors are engineered to provide the highest levels of comfort, accuracy, and precision.
Brand: Axygen™ AP-STR-KIT-P
9779.86 NOK valid until 2025-03-30
Use promo code "24134" to get your promotional price.
Description
Axygen™ Axypet™ Pro Starter Kit contains 4 variable volume pipettors in the most popular and universal volumes (0.5-10 μL, 2-20 μL, 20-200 μL, and 100-1000 μL), pipet stand for 4 pipettors, and 3 racks with tips.
- Cat. No. AP-10-P Axygen Axypet Pro Single-channel Pipettor, 0.5 to 10 μL
- Cat. No. AP-200-P Axygen Axypet Pro Single-channel Pipettor, 20 to 200 μL
- Cat. No. AP-1000-P Axygen Axypet Pro Single-channel Pipettor, 100 to 1000 μL
- Cat. No. AP-USD4-P Universal linear stand for four single-channel pipettors
- Cat. No. MR-10XT-R (10 μL) Axygen Multirack pipet tips
- Cat. No. MR-200-R (200 μL) Axygen Multirack pipet tips
- Cat. No. MR-1000XT-R (1000 μL) Axygen
Specifications
ISO8655 | |
Red/Yellow/Blue | |
1 | |
Starter Kit | |
4 single-channel pipettors, Linear stand, Sample tips, Instruction manual, QC Certificate, In-lab calibration tool | |
3 |
Autoclavable | |
Ergonomic | |
Axypet | |
Ergonomic | |
1 Kit |
Special Offers
-
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31
Days
12Hours
21Minutes
21Seconds
39 -
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
25Hours
21Minutes
21Seconds
39New Offer Save Up to 92%