missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AXIN1 Mouse anti-Human, Mouse, Rat, Clone: 1C4E8, Proteintech
Mouse Monoclonal Antibody
1696.32 NOK - 4796.50 NOK
Specifications
Antigen | AXIN1 |
---|---|
Clone | 1C4E8 |
Concentration | 1000 μg/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17619786
|
Proteintech
68093-1-IG-20UL |
20 μL |
1696.32 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17609786
|
Proteintech
68093-1-IG-150UL |
150 μL |
4796.50 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AXIN1 Monoclonal antibody apecifically detetects AXIN1 antigen in Human, Mouse, Rat Samples. It is validated for Western Blot, ELISA.Specifications
AXIN1 | |
1000 μg/mL | |
Monoclonal | |
Liquid | |
Human, Mouse, Rat | |
BC044648 | |
AXIN1 | |
IgG1 | |
Protein G purification |
1C4E8 | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
8312 | |
AXIN1 fusion protein Ag10079 | |
Primary | |
Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title