Learn More
Thermo Scientific™ Automation Tips for Caliper™ Zymark™ Liquid Handling Workstations
Increase pipetting performance with these automation tips, which undergo a rigorous 15-point quality control program to ensure accuracy and consistency.
Brand: Thermo Scientific™ 920-021
5976.29 NOK valid until 2025-12-31
Use promo code "24289" to get your promotional price.
Description
Increase pipetting performance with Thermo Scientific™ Automation Tips for Caliper™ Zymark™ Liquid Handling Workstations, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Caliper/Zymark RapidPlate, Caliper Presto, Allegro, Zephyr and SciClone.
- Available in non-sterile, sterile and with Thermo Scientific™ ART™ filter technology
- Lot-tested during production on actual workstation for which they are created
- Low percentage coefficient of variation (%CV)
Specifications
50 ÎĽL | |
Sterile | |
Rack | |
Zymark/Caliper Liquid Handling Workstations | |
Sterile and PCR Ready |
Filtered | |
Clear | |
Automation | |
Caliper Zymark |