Learn More
BRAND™ Dispenser, tilt measure without bottle
Tilt dispenser is best suited for dispensing suspensions.
Brand: BRAND™ 43066
799.03 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Calibrated to deliver (TD, Ex)
- Inscriptions in green enamel
- The automatic dispenser is particularly well-suited for dispensing suspensions
Specifications
Borosilicate Glass | |
Without bottle | |
20 mL |
Accessories and Services
Optional Accessory
-
BRAND™ Joint Clip For Tilt Measure Dispenser Joint clip for dispensers, tilt measure ... Product Code: 10044040
108.00 NOK / Each -
BRAND™ Clear Reservoir Bottle Reservoir bottle, Boro 4.1, clear1000ml Product Code: 10145680
279.00 NOK / Each -
Brand™ Ground Joint Sleeves X10 Ground joint sleeve, PTFEfor NS 29/3... Product Code: 12055332
379.00 NOK / Pack of 10
Product Content Correction
Product Title
BRAND™ Dispenser, tilt measure without bottle > Capacity (Metric): 20 mL