Learn More
Bellingham and Stanley™ Automatic Digital Polarimeter
With XPC Technology - Xylem's patented internal Peltier temperature control system
Brand:  Bellingham and Stanley™ 37-50
Product Code. 15727789
  • 180585.00 NOK / Each
Only null left
Estimated Shipment: 18-08-2025
Description
The Automatic Digital Polarimeter features a no maintenance yellow LED and interference filter. When combined with a photodiode detector these technologies allow for the reading of samples of up to 3.0 OD at the commonly used sodium (589 nm) wavelength. ADP400 polarimeters feature angular and sugar scales (ISS) and can be programmed with user scales as well as industry standard methods for displaying invert sugar, inversion (A-B) or when applying other factors such as tube length and concentration. This flexibility makes them ideal for use within laboratories where compliance with Pharmacopoeia is required.
  • Accuracy
    • ±355
    • -255 to 225 via Z sugar scale
  • Resolution
    • 0.01°A
    • 0.001°Z
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ADP450 Polarimeter
Videos
  • Bellingham and Stanley - 21 CFR Part 11 Compliance
    Video Player is loading.
    Current Time 0:00
    Duration 12:56
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 12:56
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Bellingham and Stanley - 21 CFR Part 11 Compliance
  • Bellingham and Stanley - ADP450 XPC Polarimeter 37-50
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Bellingham and Stanley - ADP450 XPC Polarimeter 37-50
    Certificates
    Promotions
    Promotions
    Product Content Correction
    Product Title
    Bellingham and Stanley™ Automatic Digital Polarimeter > Polarimeter (ADP450)