Learn More
BRAND™ Automatic Burette Fitting
Automatic Burette Fitting with micro screw, push button, and holding clamp.
Brand:  BRAND™ 23745
245.09 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10175400
  • 312.00 NOK / Each
Only null left
Estimated Shipment: 17-07-2025
Description
  • Micrometer screw with press button
  • Includes support clamp
  • For Shilling Titration Apparatus
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10 mL
Polypropylene
Buret Fitting
10 mL Automatic Burette
Suport clamp
Certificates
Promotions
Promotions
Product Content Correction
Product Title
BRAND™ Automatic Burette Fitting > Material: Polypropylene