Learn More
Abeba™ Autoclavable Clogs 39620
Antistatic and autoclavable at 135°C for 5 minutes. High slip resistance. Washing at 60°C.
Brand:  Abeba™ 39620/37
 View more versions of this product
Product Code. 15631258
  • 687.00 NOK / 1 pair
Only null left
Estimated Shipment: 25-07-2025
Description
  • CE EN ISO 20347:2012 OB A, E SRC
  • Autoclavable, sterilisable special material
  • Green
  • Slip resistance outsole sole
  • ESD dissipative according to DIN EN 61340
  • Heel strap foldable, adjustable
  • DOUBLE SIZES, please use even size when placing your order (36, 38, 40, 42, 44, 46)
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E SRC
37
39620
Green
Slip-resistant
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Product Title
Abeba™ Autoclavable Clogs 39620 > Size: 37