Learn More
3M™ Aura™ Particulate Respirator, 9300+ Gen3 Series
Click to view available options
Filter Type:
FFP2
FFP3
Assigned Protection Factor (APF):
10
20
With Exhalation Valve:
No
Yes
Quantity:
1/Pk.
10/Pk.
20/Pk.
330/Pk.
5/Pk.
Unit Size:
Pack of 10
Pack of 20
Pack of 330
Pack of 40
Pack of 5
Description
- Original 3-panel, flat-fold design fits a wide range of face shapes and sizes
- 3M™ Cool Flow™ Comfort Valve allows a greater percentage of warm, exhaled air to escape, helping to increase your comfort compared to 3M™ Aura™ 9300+ equivalent model
- 3M™ Advanced Electret (filter) Media helps you breathe easier
- Innovative tabs on the upper and lower panels simplify positioning and adjustment
- Braided headbands are colour-coded (blue for FFP2, red for FFP3) for quick and easy identification
- Embossed upper panel reduces eyewear fogging
- Easy to store away with flat-fold design and individual wrapping
Specifications
Specifications
Assigned Protection Factor (APF) | 10 |
Certifications/Compliance | CE Approved |
Certifications/Compliance (Europe) | EN 149:2001 + A1:2009, CE Approved |
Color | White |
Design Type | Flat-fold |
Disposable | Single-use |
Filter Type | FFP2 |
Material | Polypropylene |
Product Type | General Purpose Particulate Respirator |
Size | Standard |
Show More |
Documents
Scientific Resources
Brochures
3M Aura 9300PlusGen3 Brochure
File size: 3.1 MB
Flyers
3M Aura 9300PlusGen3 Flyer
File size: 1 MB
Product Content Correction
Product Title
3M™ Aura™ Particulate Respirator, 9300+ Gen3 Series >