Learn More
Attune™ Wash Solution
Brand:  Invitrogen A24974
 View more versions of this product
Product Code. 15360445
  • 383.00 NOK / 250mL
Description
Attune™ Wash Solution is an optimized cleaning buffer that is used in the Attune™ Acoustic Focusing Cytometer to maintain cleanliness of the fluidics lines, minimizing contamination or carryover.
Maintains System Cleanliness
Attune™ Wash Solution has been formulated to remove cellular debris, prevent protein building up, and minimize dyes sticking to the fluidics lines of the system. It is recommended for use during the routine maintenance and automated shutdown procedures of the Attune™ Acoustic Focusing Cytometer.
Convenient
The ready-to-use 1X wash solution was designed to eliminate the need for dilution prior to use, as well as access to a purified water system. The wash solution is stable for 30 days after the bottle is opened.
High Quality
The Attune™ Wash Solution is prepared, filtered, and packaged in a sterile environment to ensure the highest quality. Each batch is traceable by its lot number designation and has been validated for use on the Attune™ Acoustic Focusing Cytometer.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Store at room temperature.
Attune™
Wash Solution
Attune™ Acoustic Focusing Cytometer
250 mL
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3070451 Certificate TypeCertificate of Analysis Date21/01/2025 Catalog NumberA24974
Lot Number3070450 Certificate TypeCertificate of Analysis Date22/11/2024 Catalog NumberA24974
Lot Number3011768 Certificate TypeCertificate of Analysis Date18/09/2024 Catalog NumberA24974
Lot Number2970980 Certificate TypeCertificate of Analysis Date09/08/2024 Catalog NumberA24974
Lot Number2916324 Certificate TypeCertificate of Analysis Date14/06/2024 Catalog NumberA24974
Promotions
Promotions
Product Content Correction
Product Title
Attune™ Wash Solution > 250mL