missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ ATP5O (Human) Recombinant Protein
Human ATP5O full-length ORF ( AAH21233, 1 a.a. - 213 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00000539-P01.25ug
This item is not returnable.
View return policy
Description
- Sequence: MAAPAVSGLSRQVRCLSTSVVRPFAKLVRPPVQVYGIEGRYATALYSAASKQNKLEQVEKELLRVAQILKEPKVAASVLNPYVKRSIKVKSLNDITAKERFSPLTTNLINLLAENGRLSNTQGVVSAFSTMMSVHRGEVPCTVTSASPLEEATLSELKTVLKSFLSQGQVLKLEAKTDPSILGGMIVRIGEKYVDMSVKTKIQKLGRAMREIV
Specifications
AAH21233 | |
ATP synthase, H+ transporting, mitochondrial F1 complex, O subunit | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ATP5O | |
GST |
539 | |
Wheat germ expression system | |
25 μg | |
ATPO, OSCP | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |