Learn More
ASF1B Monoclonal Antibody (OTI4A4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA813137
Encompass_Preferred
 View more versions of this product
Product Code. 17577854
  • 6331.68 NOK / 100µL
Explore more special offers
Description
Description
ASF1B Monoclonal antibody specifically detects ASF1B in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
ASF1B
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9DAP7, Q9NVP2
Mouse
Protein A/G
RUO
304648, 55723, 66929
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
OTI4A4
Unconjugated
ASF1B
1700003K02Rik; AA409591; anti-silencing function 1B histone chaperone; anti-silencing function protein 1 homolog B; ASF1 anti-silencing function 1 homolog B; Asf1b; CCG1-interacting factor A-II; CIA-II; hAsf1; hAsf1b; hCIA-II; histone chaperone ASF1B; mCIA-II
Full length human recombinant protein of human ASF1B (NP_060624) produced in E.coli.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG1