Learn More
RETSCH AS 300 Standard Clamping Devices
14360.00 NOK - 201310.00 NOK
Specifications
Product Type | Clamping Devices |
---|---|
Diameter (Metric) | 315 mm |
For Use With (Equipment) | Analytical sieve shaker AS 300 |
Product Code | Brand | For Use With (Application) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | Price | Quantity & Availability | |||||
10394742
|
RETSCH
32.662.0008 |
Dry Sieving |
201310.00 NOK
Each |
Estimated Shipment: 07-04-2025
Log in to see stock. |
|||||
10172602
|
RETSCH
32.662.0012 |
Wet Sieving |
14360.00 NOK
Each |
Estimated Shipment: 16-05-2025
Log in to see stock. |
|||||
Specifications
Clamping Devices | |
315 mm | |
Analytical sieve shaker AS 300 |