missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARSB Monoclonal Antibody (OTI2A5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812952S
This item is not returnable.
View return policy
Description
Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.
ARSB Monoclonal antibody specifically detects ARSB in Human samples. It is validated for Western BlotSpecifications
ARSB | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P15848 | |
Mouse | |
Protein A/G | |
RUO | |
411 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
OTI2A5 | |
Unconjugated | |
ARSB | |
1110007C02Rik; AI480648; Arsb; Arylsulfatase B; Arylsulfatase B (MPS VI); arylsulfatase B complex; arylsulfatase B regulation; arylsulfatase B structural; arylsulfatase B temporal regulation; As1; As-1; As-1r; As1-r; As-1s; As1-s; As-1t; As1-t; ASB; Asr-1; Ast-1; G4S; MPS6; N-acetylgalactosamine-4-sulfatase | |
Human recombinant protein fragment corresponding to amino acids 84-423 of human ARSB (NP_000037) produced in E.coli. | |
30 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |