missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARL5B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11694-1-AP-20UL
This item is not returnable.
View return policy
Description
ARL8 belongs to a family of proteins that are structurally similar to ADP-ribosylation factors. ARLs and ARFs are part of the RAS superfamily of regulatory GTPases.Specifications
ARL5B | |
Polyclonal | |
Unconjugated | |
ARL5B | |
ARL5B, ARL8 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
221079, 75869 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96KC2, Q9D4P0 | |
Arl5b | |
ARL5B Fusion Protein Ag2309 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction