missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARL2BP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10090-2-AP-20UL
This item is not returnable.
View return policy
Description
ADP-ribosylation factor (ARF)-like proteins (ARLs) comprise a functionally distinct group of the ARF family of RAS-related GTPases. The protein encoded by this gene binds to ARL2. GTP with high affinity but does not interact with ARL2. GDP, activated ARF, or RHO proteins. The lack of detectable membrane association of this protein or ARL2 upon activation of ARL2 is suggestive of actions distinct from those of the ARFs. This protein is considered to be the first ARL2-specific effector identified, due to its interaction with ARL2. GTP but lack of ARL2 GTPase-activating protein activity.Specifications
ARL2BP | |
Polyclonal | |
Unconjugated | |
ARL2BP | |
ARF like 2 binding protein, ARL2BP, BART, BART1, Binder of ARF2 protein 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
107566, 23568, 498910 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q4V8C5, Q9D385, Q9Y2Y0 | |
Arl2bp | |
ARL2BP Fusion Protein Ag0138 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ARL2BP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction