Learn More
Duran™ AR Glass Disposable Culture Tubes
Disposable culture tubes for general purpose. DURAN™ AR Glass Disposable Culture Tubes are used in the laboratory setting.
Brand:  Duran™ 231721094
Product Code. 15824956
  • 384.00 NOK / Pack of 250
In Stock
to see stock.
Explore more special offers
Description
  • Made from soda-lime glass
  • With cap liner
Product Title
Duran™ AR Glass Disposable Culture Tubes > Dimensions (L x dia.): 100 x 15.75mm; Wall Thickness: 0.90mm