Learn More
Karl Hecht™ AR-Glas™ Round Bottom Centrifuge Tubes
These centrifuge tubes are suitable for a relative centrifugal force of max. 3500 x g.
584.00 NOK
Specifications
Color | Clear |
---|---|
Material | Soda Lime Glass |
For Use With | Centrifuging |
Shape | Cylindrical |
Bottom Shape | Round |
Product Code | Brand | Capacity (Metric) | Graduations | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Graduations | Price | Quantity & Availability | ||||
10013641
|
Karl Hecht™
40950016 |
10 to 12 mL |
584.00 NOK
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Manufactured from soda-lime glass
- Thick wall for durability
- Rimless
- Available with or without fused white graduations
Specifications
Clear | |
Centrifuging | |
Round | |
3500 x g |
Soda Lime Glass | |
Cylindrical | |
Centrifuge Tube |