Learn More
Aqueous-based Conductivity Standards, 1413μS/cm, Mettler Toledo™
Shop All Mettler Toledo Products
Click to view available options
Quantity:
250 mL
Specifications
Specifications
Quantity | 250 mL |
Color | Colorless |
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
New Offer Save Up to 70%
Product Content Correction
Product Title
Aqueous-based Conductivity Standards, 1413μS/cm, Mettler Toledo™ >