Learn More
Appendix IX Organophosphorous/Nitrogen Pesticides and Herbicides, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep 8270-IXJ
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
Azinphos-Methyl | 86-50-0 | 2000μg/mL |
Captafol | 2425-06-1 | 2000μg/mL |
Captan | 133-06-2 | 2000μg/mL |
Carbaryl | 63-25-2 | 2000μg/mL |
Carbofuran | 1563-66-2 | 2000μg/mL |
Coumaphos | 56-72-4 | 2000μg/mL |
Crotoxyphos | 7700-17-6 | 2000μg/mL |
Demeton O+S | 8065-48-3 | 2000μg/mL |
Dicrotophos | 141-66-2 | 2000μg/mL |
Dioxathion | 78-34-2 | 2000μg/mL |
EPN | 2104-64-5 | 2000μg/mL |
Ethion | 563-12-2 | 2000μg/mL |
Fensulfothion | 115-90-2 | 2000μg/mL |
Fenthion | 55-38-9 | 2000μg/mL |
Malathion | 121-75-5 | 2000μg/mL |
Mevinphos | 7786-34-7 | 2000μg/mL |
Monocrotophos | 6923-22-4 | 2000μg/mL |
Octamethylpyrophosphoramide | 152-16-9 | 2000μg/mL |
Phosalone | 2310-17-0 | 2000μg/mL |
Phosphamidon(33.4% E; 64.6% Z) | 13171-21-6 | 2000μg/mL |
Terbufos | 13071-79-9 | 2000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Appendix IX Organophosphorus/Nitrogen Pest & Herb | |
2mL amber prescored glass ampule | |
For US EPA Methods 8270, 8270C | |
Acetone |
Method 8270 | |
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. | |
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation | |
1 mL |
Safety and Handling
- Appendix IX Organophosphorus/Nitrogen Pest & Herb
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Carcinogenicity Category 1B
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H350-May cause cancer.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : acetone, captafol, captan, carbaryl, malathion, azinphos-methyl, phosalone
Product intended for laboratory use only