missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apc1 Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-86985AF700
This item is not returnable.
View return policy
Description
Apc1 Polyclonal antibody specifically detects Apc1 in Human, Mouse samples. It is validated for Western Blot
Specifications
Apc1 | |
Polyclonal | |
Western Blot | |
anaphase promoting complex subunit 1, anaphase-promoting complex 1 (meiotic checkpoint regulator), anaphase-promoting complex subunit 1, APC1Testis-specific gene 24 protein, Cyclosome subunit 1, Mitotic checkpoint regulator, TSG24MCPR | |
The immunogen recognized by this antibody maps to a region between residue 500 and 550 of human anaphase promoting complex subunit 1 using the numbering given in entry NP_073153.1 (GeneID 64682). | |
0.1 mL | |
Cell Biology, Cell Cycle and Replication, Phospho Specific, Ubiquitin Proteasome Pathway | |
64682 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Apc1 Antibody, Alexa Fluor™ 700, Novus Biologicals™ > 0.1 mL; Alexa Fluor 700
Spot an opportunity for improvement?Share a Content Correction