Learn More
Invitrogen™ Anza™ 91 Acc65I
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
Brand: Invitrogen™ IVGN0916
Includes: Anza™ Restriction Enzyme, Anza™ 10X Buffer, Anza™ 10X Red Buffer
Description
Simplify traditional restriction enzyme cloning with the Invitrogen™ Anza™ Restriction Enzyme Cloning System.
The Invitrogen Anza Restriction Enzyme Cloning System utilizes a single buffer system for complete digestions and incorporates Anza cloning enzymes into a clear and simple system.- One buffer for all restriction enzymes
- One digestion protocol for all DNA types
- Complete digestion in 15 min
- Overnight digestion without star activity
Certifications
CoA

Specifications
300 μL Anza 91 Acc65I 1 mL Anza Buffer (10X) 1 mL Anza Red Buffer (10X) Store at -20°C. |
|
CpG Methylation-Sensitive, Not CpG Methylation-Sensitive, Not Dam Methylation-Sensitive | |
Anza™ 10X Red Buffer, Anza™ 10X Buffer | |
37°C | |
1500 units | |
Traditional Cloning |
Approved for shipment on Wet or Dry Ice | |
Acc65I | |
Yes | |
No | |
Restriction Enzyme Cloning System | |
Anza™ |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2883886 | Certificate of Analysis | 28/01/2025 | IVGN0916 |
2987899 | Certificate of Analysis | 26/01/2025 | IVGN0916 |
2996371 | Certificate of Analysis | 26/01/2025 | IVGN0916 |
3033516 | Certificate of Analysis | 26/01/2025 | IVGN0916 |
3071852 | Certificate of Analysis | 25/01/2025 | IVGN0916 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.