Learn More
anti-TIMP1, Human, Matched Antibody Pair, Abnova™
To detect and quantify protein level of human
Brand: Abnova H00007076-AP41.1Set
Includes: Capture antibody: mouse monoclonal anti-TIMP1 (100μg) , Detection antibody: biotinylated mouse monoclonal anti-TIMP1, IgG1 Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Specifications
TIMP1 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human TIMP1. | |
CLGI/EPA/EPO/FLJ90373/HCI/TIMP | |
Mouse | |
RUO | |
7076 | |
Human | |
Liquid |
ELISA | |
Biotin | |
TIMP1 | |
TIMP1 | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-TIMP1 (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-TIMP1, IgG1 Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
For Research Use Only