Learn More
Zwiebel™ Anti-static Brush
Clean Zwiebel™ weights with these anti-static Brushes.
Brand:  Zwiebel™ PBI2528A
Product Code. 11765045
  • 259.00 NOK / Each
Only null left
Estimated Shipment: 16-07-2025
Edge
Add to basket
Description
Description
  • Flat synthetic fiber brush
  • Intended for the superficial cleaning of Zwiebel weights
Specifications
Specifications
Brush
185 mm
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Zwiebel™ Anti-static Brush > Length: 185mm