missing translation for 'onlineSavingsMsg'
Learn More

Oct3/4 Mouse, PE, Clone: 40/OCT-3, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 560186

3205.72 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 15849998

  • 3210.00 NOK / Each

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Development of a multicellular organism from a single fertilized cell is regulated by the coordinated activity of DNA transcription factors. Oct3/4, a member of the POU family of transcription factors, functions in pluripotent cells of early embryonic stem cell lines and embryonal carcinomas. Other members of the POU family include Oct1, Oct2, Pit-1, and unc-86. The POU domain, a 150-amino acid region that determines binding specificity, is conserved among these proteins and consists of 3 subdomains: POU-specific A and B subdomains and a homeobox-like subdomain. Oct3/4 is expressed in undifferentiated cells, but is lost as cells are induced to differentiate. Oct3/4 is not expressed in adult tissues. The interaction of Oct3/4 with SOX2, another embryonic transcription factor, produces an active complex that regulates expression of genes such as Nanog, UTF1, and FGF4. Although Oct3/4 is specifically phosphorylated on serine residues, this modification is not required for DNA binding, but may affect its transactivation potential. Thus, Oct3/4 is a transcription factor that plays an important role in determining early steps of embryogenesis and differentiation.

Host Species: Mouse
Clone: 40
Isotype: IgG1 Îş
Species Reactivity: Human
Immunogen: Mouse Oct3 aa. 252-372 Recombinant Protein

Intracellular Staining

TRUSTED_SUSTAINABILITY
Specifications

Specifications

OCT3/4
Monoclonal
PE
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse Oct3 aa. 252-372 Recombinant Protein
50 Tests
Cell Biology
Human, Mouse
IgG1 Îş
Flow Cytometry
40/OCT-3
Oct3, OTF3, Oct4, OTF4, POU5F1
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Oct3/4 Mouse, PE, Clone: 40/OCT-3, BD > 50 Tests; PE

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.