missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-NHLH2, Human, Matched Antibody Pair, Abnova™
To detect and quantify protein level of human
Brand: Abnova H00004808-AP41.1Set
Includes: Capture antibody: mouse monoclonal anti-NHLH2, IgG2a Kappa (100μg) , Detection antibody: biotinylated mouse monoclonal anti-NHLH2, IgG2a Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Specifications
NHLH2 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human NHLH2. | |
HEN2/KIAA0490/NSCL2/bHLHa34 | |
Mouse | |
RUO | |
4808 | |
Human | |
Liquid |
ELISA | |
Biotin | |
NHLH2 | |
NHLH2 | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-NHLH2, IgG2a Kappa (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-NHLH2, IgG2a Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
For Research Use Only