Learn More
anti-Mouse CX3CR1, APC, Polyclonal, R&D Systems™
Goat Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems FAB5825A-025

Description
CX3CR1 Polyclonal specifically detects CX3CR1 in Mouse samples. It is validated for Flow Cytometry.Specifications
CX3CR1 | |
Polyclonal | |
Flow Cytometry 10 uL/10^6 cells | |
Q9Z0D9 | |
CX3CR1 | |
E. coli-derived recombinant mouse CX3CR1 Met1-Thr32, Leu92-Lys104, Thr169-Val196, Lys258-Leu274 Accession # Q9Z0D9 | |
25 Tests | |
1524 | |
Mouse | |
IgG |
Flow Cytometry | |
APC | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
Beta chemokine receptor-like 1, CCRL1, chemokine (C-C) receptor-like 1, chemokine (C-X3-C motif) receptor 1, chemokine (C-X3-C) receptor 1, CMKbRL1, CMK-BRL1, CMK-BRL-1, CMKDR1, CX3C chemokine receptor 1, Fractalkine receptor, G protein-coupled receptor 13, GPR13G-protein coupled receptor 13, GPRV28, V28, V28C-X3-C CKR-1 | |
Goat | |
Affinity Purified | |
Primary | |
Detects mouse CX3CR1 in direct ELISAs. Stains mouse CX3CR1 transfectants but not irrelevant transfectants in flow cytometry. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.