Learn More
anti-IL8, Human, Matched Antibody Pair, Abnova™
To detect and quantify protein level of human
Brand:  Abnova H00003576-AP41.1Set
Product Code. 16152244
  • 9950.00 NOK / 1 set
Explore more special offers
Includes: Capture antibody: mouse monoclonal anti-IL8, IgG2a Kappa (100μg) , Detection antibody: biotinylated mouse monoclonal anti-IL8, IgG2a Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Specifications
IL8
Monoclonal/Polyclonal
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human IL8.
CXCL8/GCP-1/GCP1/LECT/LUCT/LYNAP/MDNCF/MONAP/NAF/NAP-1/NAP1
Mouse
RUO
3576
Human
Liquid
ELISA
Biotin
IL8
IL8
1 Set
Primary
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-IL8, IgG2a Kappa (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-IL8, IgG2a Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.