missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ErbB3 Monoclonal Antibody (2B5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA513040
This item is not returnable.
View return policy
Description
MA5-13040 targets HER-3 in IP and WB applications and shows reactivity with Human, mouse, and Rat samples. The MA5-13040 immunogen is recombinant rat c-erbB-3/HER-3 oncoprotein.
ErbB3 Monoclonal antibody specifically detects ErbB3 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blot.Specifications
ErbB3 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P21860, Q61526, Q62799 | |
ERBB3 | |
Recombinant rat c-erbB-3/HER-3 oncoprotein | |
500 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
2B5 | |
Unconjugated | |
ERBB3 | |
avian erythroblastosis oncogene B 3; avian erythroblastosis oncogene B 3 receptor; C76256; c-erbB3; c-erbB-3; C-errB-3; erb-b2 receptor tyrosine kinase 3; ERBB3; Erbb-3; Erbb3r; erbB3-S; glial growth factor receptor; HER3; human epidermal growth factor receptor 3; LCCS2; MDA BF 1; MDA-BF-1; MGC88033; nuc-ErbB3; p180 ErbB3; p180-ErbB3; p45 sErbB3; p45-sErbB3; p85 sErbB3; p85-sErbB3; proto-oncogene-like protein c-ErbB-3; receptor tyrosine kinase; receptor tyrosine-protein kinase erbB-3; receptor tyrosine-protein kinase erbB-3; LOW QUALITY PROTEIN: receptor tyrosine-protein kinase erbB-3; tyrosine kinase-type cell surface receptor HER3; v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 3; v-erb-b2 erythroblastic leukemia viral oncogene homolog 3; v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian) | |
Mouse | |
Protein G | |
RUO | |
13867, 2065, 29496 | |
4° C | |
Liquid |